missing translation for 'onlineSavingsMsg'
Learn More

Tamibarotene, MedChemExpress

Product Code. 16472257
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472257 50 mg 50mg
16406128 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16472257 Supplier Medchem Express Supplier No. HY14652
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tamibarotene is a retinoic acid receptor α/β (RARα/β) agonist, showing high selectivity over RARγ.

Tamibarotene is a retinoic acid receptor α/β (RARα/β) agonist, showing high selectivity over RARγ.

Specifications

Chemical Name or Material Tamibarotene
CAS 94497-51-5
Color White
Purity Grade Notes Research
Molecular Formula C22H25NO3
Quantity 50 mg
Synonym Am 80
Solubility Information DMSO : 25.5 mg/mL (72.56 mM; Need ultrasonic and warming)
SMILES O=C(C1=CC=C(C=C1)C(NC2=CC=C3C(C)(CCC(C)(C3=C2)C)C)=O)O
Molecular Weight (g/mol) 351.44
Formula Weight 351.44
Percent Purity 98.66%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.