missing translation for 'onlineSavingsMsg'
Learn More

Taltirelin, Tocris Bioscience™

Synthetic thyrotropin-releasing hormone analog

Specifications

CAS 103300-74-9
Content And Storage Store at +4°C
View More Specs

Description

Description

Synthetic thyrotropin-releasing hormone (TRH) analog. Displays ∽ 30 - 100-fold more potent CNS activity and ∽ 50-fold weaker endocrine activity than TRH. Antinociceptive and neuroprotective.
Specifications

Specifications

103300-74-9
Store at +4°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.