missing translation for 'onlineSavingsMsg'
Learn More

Talmapimod, MedChemExpress

Product Code. 16443948
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443948 10 mg 10mg
16432926 25 mg 25mg
16426667 5 mg 5mg
16400307 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16443948 Supplier Medchem Express Supplier No. HY10406

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Talmapimod (SCIO-469) is an orally active, selective, and ATP-competitive p38α inhibitor with an IC50 of 9 nM. Talmapimod shows about 10-fold selectivity over p38β, and at least 2000-fold selectivity over a panel of 20 other kinases, including other MAPKs.

Talmapimod (SCIO-469) is an orally active, selective, and ATP-competitive p38α inhibitor with an IC50 of 9 nM. Talmapimod shows about 10-fold selectivity over p38β, and at least 2000-fold selectivity over a panel of 20 other kinases, including other MAPKs.

Specifications

Chemical Name or Material Talmapimod
CAS 309913-83-5
Color Pink
Purity Grade Notes Research
Molecular Formula C27H30ClFN4O3
Quantity 10 mg
Synonym SCIO-469
Solubility Information DMSO : ≥ 100 mg/mL (194.93 mM)
SMILES O=C(N(C)C)C(C1=CN(C)C2=C1C=C(C(N3[C@H](C)CN(CC4=CC=C(F)C=C4)[C@@H](C)C3)=O)C(Cl)=C2)=O
Molecular Weight (g/mol) 513.0
Formula Weight 513.0
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.