Learn More
Köttermann Tall Safety Storage Cabinet with Doors
This Tall Safety Storage Cabinet offers top safety for flammable hazardous substances, complying with fire resistance class standards (Type 90).
Brand:  Köttermann 371-00030
Additional Details : Weight : 433.00000kg
 View more versions of this product
Product Code. 17372129
  • £4181.70 / Each
Estimated Shipment: 21-08-2024
to see stock.
Description
Description
  • Different versions that meet your needs
  • Additional storage space
  • Top safety for flammable hazardous substances (Type 90)
Specifications
Specifications
1 Drip Tray
Hinged, Auto-close
2
615 x 1200 x 1955 mm
6