missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TALK-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144235
This item is not returnable.
View return policy
Description
TALK-1 Polyclonal Antibody for IHC (P), ELISA
The KCNK16 gene encodes the TALK-1 channel, an abundant K+ channel transcript found in mouse and in human Beta-cells. There are four human TALK-1 transcript variants, including two that form functional K+ channels (TALK-1a (transcript variant 2) and TALK1-b (transcript variant 3). The TALK-1 structure includes 4 transmembrane domains, 2 extracellular loops, and intracellular N- and C-termini. The C-terminal tail of TALK-1 is a critical component that influences the magnitude of TALK-1 channel activation, while the N-terminus is important for protein interaction. TALK-1, two-pore domain K+ (K2p 16.1) channel is responsible for modulating glucose stimulated insulin secretion that relies on Beta-cell CA2+ influx. The channel produces outwardly rectifying, non-inactivating K+ currents, which are enhanced by elevations in extracellular pH. TALK-1 is thus pH-sensitive, with increased activity under alkaline conditions. In addition, it can be activated by singlet oxygen and nitric oxide.
Specifications
| TALK-1 | |
| Polyclonal | |
| Unconjugated | |
| KCNK16 | |
| 2P domain potassium channel Talk-1; 4731413G05Rik; K2p16.1; KCNK16; pancreatic potassium channel Talk-1; potassium channel subfamily K member 16; potassium channel, subfamily K, member 16; potassium channel, two pore domain subfamily K, member 16; potassium two pore domain channel subfamily K member 16; TALK1; TALK-1; TWIK-related alkaline pH-activated K(+) channel 1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 83795 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 3.5 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q96T55 | |
| KCNK16 | |
| Recombinant Human Potassium channel subfamily K member 16 protein (259-309AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction