missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talin1 Antibody (TA205), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-21642AF750
This item is not returnable.
View return policy
Description
Talin1 Monoclonal specifically detects Talin1 in Human, Chicken, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| Talin1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TLN1 | |
| Human platelet talin. | |
| Primary | |
| Immunoblots the 225 kDa talin band on SDS gels of platelets and other cells. It may be used to stain adhesion plaques in fixed and permeabilized fibroblasts. TA205 recognizes an epitope within residues 139-433. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| TA205 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| ILWEQ, KIAA1027, talin 1, talin-1, TLN | |
| Mouse | |
| 0.1 mL | |
| 7094 | |
| Human, Chicken, Rabbit | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction