missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talin1 Antibody (97H6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£169.00 - £336.00
Specifications
| Antigen | Talin1 |
|---|---|
| Clone | 97H6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633109
|
Novus Biologicals
NBP2-50320-0.025ml |
0.025 mL |
£169.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624578
|
Novus Biologicals
NBP2-50320 |
0.1 mL |
£336.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Talin1 Monoclonal antibody specifically detects Talin1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| Talin1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers | |
| PBS | |
| 7094 | |
| IgG1 | |
| Protein A purified |
| 97H6 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ILWEQ, KIAA1027, talin 1, talin-1, TLN | |
| His-tagged polypeptide containing residues 489-911 of mouse Talin1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title