missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Talin Monoclonal Antibody (TA205)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528133
This item is not returnable.
View return policy
Description
Mouse anti Human Talin antibody, clone TA205 recognizes human talin, a approximately 225 kDa cytoskeletal protein.
Talin, a multifunctional constituent of cell-substratum attachment sites, is a high molecular weight protein (225-235 kDa) found in variety of tissues and cell types. It is localized at a subset of adherens junctions, specialized cell-cell and cell-matrix associations that are characterized by the presence of filamentous actin at the cytoplasmic face of the junctional complex. In cultured cells, talin is absent from cell-cell junctions and found predominantly at adhesion plaques and in fibrillar streaks underlying cell surface fibronectin. Talin interacts with at least two other proteins that are localized at adhesion plaques, vinculin and integrin.
Specifications
| Talin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q9Y490, Q9Y4G6 | |
| TLN1, TLN2 | |
| Human platelet talin. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| TA205 | |
| Unconjugated | |
| TLN2 | |
| 5730421P04Rik; AI507121; AI787438; AL118320; ILWEQ; KIAA0320; KIAA1027; mKIAA0320; RGD1565416; talin 1; talin 2; talin-1; Talin-2; TLN; Tln1; Tln2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 7094, 83660 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction