missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Talin 2 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148892
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a protein related to talin 1, a cytoskeletal protein that plays a significant role in the assembly of actin filaments and in spreading and migration of various cell types, including fibroblasts and osteoclasts. This protein has a different pattern of expression compared to talin 1 but, like talin 1, is thought to associate with unique transmembrane receptors to form novel linkages between extracellular matrices and the actin cytoskeleton.
Specifications
| Talin 2 | |
| Polyclonal | |
| Unconjugated | |
| Tln2 | |
| 5730421P04Rik; AI507121; AI787438; AL118320; ILWEQ; KIAA0320; mKIAA0320; RGD1565416; talin 2; talin-2; TLN2 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 83660 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9Y4G6 | |
| Tln2 | |
| E. coli-derived recombinant human Talin2, Ala977-Arg1183. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction