Learn More
Invitrogen™ Talin 2 Monoclonal Antibody (8d4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549310
Description
Adding 1 mL of PBS buffer will yield a concentration of 100 μg/mL. Positive Control - WB: human Daudi whole cell, mouse NIH/3T3 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a protein related to talin 1, a cytoskeletal protein that plays a significant role in the assembly of actin filaments and in spreading and migration of various cell types, including fibroblasts and osteoclasts. This protein has a different pattern of expression compared to talin 1 but, like talin 1, is thought to associate with unique transmembrane receptors to form novel linkages between extracellular matrices and the actin cytoskeleton.
Specifications
| Talin 2 | |
| Monoclonal | |
| 100 μg/mL | |
| Ascites, 1.2% sodium acetate with 2mg BSA and 0.01mg sodium azide | |
| Q71LX4, Q9Y4G6 | |
| Tln2 | |
| Purified chicken gizzard talin. | |
| RUO | |
| 315776, 415374, 70549, 83660 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 8d4 | |
| Unconjugated | |
| Tln2 | |
| 5730421P04Rik; AI507121; AI787438; AL118320; ILWEQ; KIAA0320; mKIAA0320; RGD1565416; talin 2; talin-2; TLN2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Chicken, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.