Learn More
Talin-1 Rabbit, anti-Human, Clone: 7D11, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82856-4-RR-20UL
Description
The Talin-1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Talin-1. This antibody recognizes human, mouse, rat antigen. The Talin-1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| Talin-1 | |
| 7D11 | |
| ILWEQ, KIAA1027, talin 1, TLN, TLN1 | |
| 20 μL | |
| 7094 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunofluorescence, ELISA | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |