Learn More
Invitrogen™ Talin 1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA580133
Product Code. 15965695
  • £393.00 / 100µg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human HepG2 whole cell, human Jurkat whole cell, rat C6 whole cell, mouse NIH/3T3 whole cell. IHC: human Lung cancer tissue, mouse small intestine tissue, human intestinal cancer tissue, human tonsil tissue, rat small intestine tissue, rat lung tissue, rat spleen tissue, mouse spleen tissue.
Talin, a multifunctional constituent of cell-substratum attachment sites, is a high molecular weight protein (225-235 kDa) found in variety of tissues and cell types. It is localized at a subset of adherens junctions, specialized cell-cell and cell-matrix associations that are characterized by the presence of filamentous actin at the cytoplasmic face of the junctional complex. In cultured cells, talin is absent from cell-cell junctions and found predominantly at adhesion plaques and in fibrillar streaks underlying cell surface fibronectin. Talin interacts with at least two other proteins that are localized at adhesion plaques, vinculin and integrin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Talin 1
Polyclonal
Unconjugated
TLN1
ILWEQ; KIAA1027; talin 1; talin-1; TLN; TLN1
Rabbit
Antigen affinity chromatography
RUO
21894, 313494, 7094
-20°C
Lyophilized
Immunohistochemistry (Paraffin), Western Blot
500 μg/mL
PBS with 4mg trehalose and 0.05mg sodium azide
P26039, Q9Y490
TLN1
A synthetic peptide corresponding to a sequence of human Talin 1 (QQLAPREGISQEALHTQMLTAVQEISHLIEPLANAARAEASQ).
100 μg
Primary
Human, Mouse, Rat