missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Talin 1 Monoclonal Antibody (GT24212)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527787
This item is not returnable.
View return policy
Description
Positive Control: H1299 Predicted Reactivity: Mouse (99%), Rat (99%), Zebrafish (92%), Chicken (93%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Talin, a multifunctional constituent of cell-substratum attachment sites, is a high molecular weight protein (225-235 kDa) found in variety of tissues and cell types. It is localized at a subset of adherens junctions, specialized cell-cell and cell-matrix associations that are characterized by the presence of filamentous actin at the cytoplasmic face of the junctional complex. In cultured cells, talin is absent from cell-cell junctions and found predominantly at adhesion plaques and in fibrillar streaks underlying cell surface fibronectin. Talin interacts with at least two other proteins that are localized at adhesion plaques, vinculin and integrin.
Specifications
| Talin 1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9Y490 | |
| TLN1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human Talin-1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| GT24212 | |
| Unconjugated | |
| TLN1 | |
| ILWEQ; KIAA1027; talin 1; talin-1; TLN; TLN1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 313494, 7094 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction