Learn More
Invitrogen™ Tali™ Cellular Analysis Slides
For use with Tali™ Image-Based Cytometer
Brand: Invitrogen™ T10795
2242.00 GBP valid until 2024-12-31
Use promo code "21956" to get your promotional price.
Code : 50
Additional Details : Weight : 0.80000kg
Description
The TaliCellular Analysis Slides are specifically designed for use with the TaliImage-Based Cytometer. Each slide contains two separate, enclosed analysis chambers designed to accommodate 25μL of sample per chamber.
- Each TaliCellular Analysis slide is pre-sterilized and individually packaged
- Since TaliImage-Based Cytometer is fluorescent cellular analysis instrument slides are composed of low fluorescent plastic
- Slides are disposable
- After use, slides can be disposed of with regular biohazard waste
Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cellular Imaging, Imaging Cytometry
Specifications
Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cellular Imaging, Imaging Cytometry | |
Contains ten boxes of 50 slides | |
2°C to 25°C | |
500 Slides |
For Research Use Only. Not for use in diagnostic procedures.