missing translation for 'onlineSavingsMsg'
Learn More

Talc, Pure, Fine Powder, Purified By Acids, Fisher Chemical

Catalog Number 10687612
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy
Melting Point 800°C
CAS 14807-96-6
Color White
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula H2Mg3O12Si4
MDL Number MFCD00792903
Synonym magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate
InChI Key KNXVOGGZOFOROK-UHFFFAOYSA-N
SMILES [Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O
IUPAC Name dioxosilane;oxomagnesium;hydrate
Molecular Weight (g/mol) 379.26
PubChem CID 16211421
Formula Weight 379.28g/mol
Packaging Plastic powder jar
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'