missing translation for 'onlineSavingsMsg'
Learn More

Talarozole, MedChemExpress

Product Code. p-200098870
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496476

Brand: Medchem Express HY14531

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Talarozole (R115866) is an oral systemic all-trans retinoic acid metabolism blocking agent (RAMBA) which increases intracellular levels of endogenous all-trans retinoic acid (RA). Talarozole inhibits both CYP26A1 and CYP26B1 with IC50s of 5.4 and 0.46 nM, respectively.

Talarozole (R115866) is an oral systemic all-trans retinoic acid metabolism blocking agent (RAMBA) which increases intracellular levels of endogenous all-trans retinoic acid (RA). Talarozole inhibits both CYP26A1 and CYP26B1 with IC50s of 5.4 and 0.46 nM, respectively.

Specifications

Chemical Name or Material Talarozole
CAS 201410-53-9
Color White
Purity Grade Notes Research
Molecular Formula C21H23N5S
Quantity 100 mg
Synonym R115866
Solubility Information DMSO : ≥ 36 mg/mL (95.36 mM)
SMILES CCC(CC)C(C1=CC=C(NC2=NC3=CC=CC=C3S2)C=C1)N4N=CN=C4
Molecular Weight (g/mol) 377.51
Formula Weight 377.51
Percent Purity 98.53%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.