missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Talabostat Mesylate

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Talabostat mesylate is a potent DPP IV inhibitor (IC50 values are <4, 4, 11, 310 and 560 nM at DPP4, DPP8, DPP9, DPP7 and FAP, respectively). Activates the inflammasome sensor protein NLRP1b and selectively triggers pyroptosis in monocytes and macrophages. Induces immune-mediated tumor regression in multiple mouse models of cancer. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | B-[(2R)-1-[(2S)-2-Amino-3-methyl-1-oxobutyl]-2-pyrrolidinyl]boronic acid methanesulfonate |
| CAS | 150080-09-4 |
| Synonym | Val-boroPro, BXCL 701 |
| Quantity | 10 mg |
| Molecular Formula | C9H19BN2O3.CH3SO3H |
| Formula Weight | 310.18 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction