missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530586
This item is not returnable.
View return policy
Description
Recommended positive controls: H1299. Predicted reactivity: Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TAL-1 is a basic helix-loop-helix transcription. It regulates differentiation and survival during hemopoiesis. TAL-1 is also implicated in the genesis of hemopoietic malignancies and it may play an important role in hemopoietic differentiation. It serves as a positive regulator of eryhtroid differentiation. Mutations are associated with T-cell leukemia and melanoma. TAL-1 is know to bind to the LIM domain containing protein Rhombotin-2.
Specifications
| TAL1 | |
| Polyclonal | |
| Unconjugated | |
| TAL1 | |
| basic helix-loop-helix transcription factor; bHLHa17; Class A basic helix-loop-helix protein 17; HGNC:11556; LOW QUALITY PROTEIN: T-cell acute lymphocytic leukemia protein 1; Scl; scl/tal1; SCL/tal-1; SCL-alpha; SCL-beta; si:ch211-135j1.3; stem cell leukemia protein; stem cell leukemia protein SCL; stem cell protein; stem-cell leukemia hematopoietic transcription factor; T cell acute lymphocytic leukemia 1; TAL bHLH transcription factor 1, erythroid differentiation factor; TAL1; Tal-1; tal-1 product; T-cell acute lymphocytic leukemia 1; T-cell acute lymphocytic leukemia protein 1; T-cell acute lymphocytic leukemia protein 1 homolog; T-cell leukemia/lymphoma protein 5; TCL5; transcription factor; zgc:85605; zSCL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 21349, 6886 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1.61 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P17542, P22091 | |
| TAL1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human TAL1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction