missing translation for 'onlineSavingsMsg'
Learn More

Takinib, MedChemExpress

Product Code. 16460227
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16460227

Brand: Medchem Express HY103490

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Takinib (EDHS-206) is an orally active and selective TAK1 inhibitor (IC50=9.5 nM), more than 1.5 log more potent than the second and third ranked targets, IRAK4 (120 nM) and IRAK1 (390 nM), respectively.

Takinib is an inhibitor of autophosphorylated TAK1 that non-competitively binds within the ATP binding pocket. Takinib induces apoptosis following TNFα stimulation in cell models of rheumatoid arthritis and metastatic breast cancer. Takinib is also a P. falciparum protein kinase 9 (PfPK9) inhibitor (KD(app) of 0.46 μM).

Specifications

Chemical Name or Material Takinib
Physical Form Solid
CAS 1111556-37-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H18N4O2
For Use With (Application) Cancer-Kinase/protease
Synonym EDHS-206
Solubility Information DMSO : 50 mg/mL (155.11 mM; Need ultrasonic)
SMILES CCCN1C2=CC=CC=C2N=C1NC(C3=CC(C(N)=O)=CC=C3)=O
Molecular Weight (g/mol) 322.36
Formula Weight 322.36
Percent Purity 99.15%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.