missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Takinib

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
Takinib is a potent and selective TAK1/MAP3K7 kinase inhibitor (IC50 = 8.2-9.5 nM). Selective for TAK1 over IRAK4, IRAK1, GCK, CLK2 and MINK1 (IC50 values = 120, 390, 430, 430 and 1900 nM, respectively). Demonstrates weak inhibition at Src and Yes1. Exhibits no inhibition of MAP2K or other MAP3K family members. Induces apoptosis following TNF-α stimulation in in vitro models of rheumatoid arthritis and metastatic breast cancer.
Specifications
Specifications
| Chemical Name or Material | N1-(1-Propyl-1 H-benzimidazol-2-yl)-1,3-benzenedicarboxamide |
| CAS | 1111556-37-6 |
| Quantity | 5 mg |
| Target | Other Kinase Inhibitors |
| Molecular Formula | C18H18N4O2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction