missing translation for 'onlineSavingsMsg'
Learn More

TAK1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18348003 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348003 100 μg 100µL
18389304 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18348003 Supplier Novus Biologicals Supplier No. NBP316950100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TAK1 Polyclonal antibody specifically detects TAK1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 50% glycerol, pH7.3
Gene Alias EC 2.7.11, EC 2.7.11.25, MEKK7, mitogen-activated protein kinase kinase kinase 7, TAK1/MAP3K7, TAK1TGF1a, TGF-beta activated kinase 1, TGF-beta-activated kinase 1, Transforming growth factor-beta-activated kinase 1
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around T187 of human TAK1 (NP_663304.1). HMTNN
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6885
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.