missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAK-960, MedChemExpress
Description
TAK-960 is an orally available, selective inhibitor of polo-like kinase 1 (PLK1), with an IC50 of 0.8 nM. TAK-960 also shows inhibitory activities against PLK2 and PLK3, with IC50s of 16.9 and 50.2 nM, respectively. TAK-960 inhibits proliferation of multiple cancer cell lines and exhibits significant efficacy against multiple tumor xenografts.
Specifications
Specifications
| Chemical Name or Material | TAK-960 |
| CAS | 1137868-52-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C27H34F3N7O3 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 16.67 mg/mL (29.68 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(NC1CCN(C)CC1)C2=CC(OC)=C(NC3=NC=C(N4C)C(N(C5CCCC5)CC(F)(F)C4=O)=N3)C=C2F |
| Molecular Weight (g/mol) | 561.6 |
| Formula Weight | 561.6 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?