missing translation for 'onlineSavingsMsg'
Learn More

TAK-875, TRC

Product Code. 17953725
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17953725 5 mg 5mg
17903685 25 mg 25mg
17943945 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17953725 Supplier Toronto Research Chemicals Supplier No. TRCT0049155MG
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1000413-72-8
Molecular Formula C29 H32 O7 S
Molecular Weight (g/mol) 524.6252
Synonym (3S)-6-[[2',6'-Dimethyl-4'-[3-(methylsulfonyl)propoxy][1,1'-biphenyl]-3-yl]methoxy]-2,3-dihydro-3-benzofuranacetic Acid, (+)-TAK 875, TAK-875
IUPAC Name 2-[(3S)-6-[[3-[2,6-dimethyl-4-(3-methylsulfonylpropoxy)phenyl]phenyl]methoxy]-2,3-dihydro-1-benzofuran-3-yl]acetic acid
SMILES Cc1cc(OCCCS(=O)(=O)C)cc(C)c1c2cccc(COc3ccc4[C@H](CC(=O)O)COc4c3)c2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C29H32O7S/c1-19-12-25(34-10-5-11-37(3,32)33)13-20(2)29(19)22-7-4-6-21(14-22)17-35-24-8-9-26-23(15-28(30)31)18-36-27(26)16-24/h4,6-9,12-14,16,23H,5,10-11,15,17-18H2,1-3H3,(H,30,31)/t23-/m1/s1
Formula Weight 524.1869
Chemical Name or Material 2-[(3S)-6-[(3-[2,6-dimethyl-4-(3-methylsulfonylpropoxy)phenyl]phenyl)methoxy]-2,3-dihydro-1-benzofuran-3-yl]acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.