missing translation for 'onlineSavingsMsg'
Learn More

TAK-779, MedChemExpress

Product Code. 16488357
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16488357

Brand: Medchem Express HY13406

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TAK-779 is a potent and selective nonpeptide antagonist of CCR5 and CXCR3, with a Ki of 1.1 nM for CCR5, and effectively and selectively inhibits R5 HIV-1, with EC50 and EC90 of 1.2 nM and 5.7 nM, respectively, in MAGI-CCR5 cells.

TAK-779 is a potent and selective nonpeptide antagonist of CCR5 and CXCR3, with a Ki of 1.1 nM for CCR5, and effectively and selectively inhibits R5 HIV-1, with EC50 and EC90 of 1.2 nM and 5.7 nM, respectively, in MAGI-CCR5 cells.

Specifications

Chemical Name or Material TAK-779
Physical Form Solid
CAS 229005-80-5
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C33H39ClN2O2
For Use With (Application) COVID-19-anti-virus
Synonym Takeda 779
Solubility Information DMSO : ≥ 25 mg/mL (47.07 mM) ∣H2O : 16.66 mg/mL (31.37 mM; Need ultrasonic and warming)
SMILES C[N+](C)(CC1=CC=C(NC(C2=CC3=CC(C4=CC=C(C)C=C4)=CC=C3CCC2)=O)C=C1)C5CCOCC5.[Cl-]
Molecular Weight (g/mol) 531.13
Formula Weight 531.13
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.