missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TAK 779
GREENER_CHOICE

Product Code. 18727196
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18727196

Brand: Tocris Bioscience™ 4340/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective CCR5 and CCR2 chemokine receptor antagonist

TAK 779 is a potent and selective CCR5 and CCR2 chemokine receptor antagonist (IC50 values = 1 nM and 27 nM, respectively). TAK 779 blocks the binding of macrophage inflammatory protein 1α and 1β to CHO cells (IC50 = 1 nM) and inhibits replication of R5 HIV-1 at 1.6-3.7 nM. In animal models, TAK 779 protects against ischemic brain injury and prevents development of asthma features.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[[4-[[[6,7-Dihydro-2-(4-methylphenyl)-5 H-benzocyclohepten-8-yl]carbonyl]amino]phenyl]methyl]tetrahydro-N,N-dimethyl-2 H-pyran-4-aminium chloride
CAS 229005-80-5
Quantity 10 mg
Target Chemokine Receptor Antagonists
Molecular Formula C33H39N2O2.Cl
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.