missing translation for 'onlineSavingsMsg'
Learn More

TAK-580, MedChemExpress

Product Code. p-200097471
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16466686

Brand: Medchem Express HY15246

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TAK-580 (MLN 2480) is an orally active and selective inhibitor of pan-Raf kinase.

TAK-580 (MLN 2480) is an orally active and selective inhibitor of pan-Raf kinase.

Specifications

Chemical Name or Material TAK-580
CAS 1096708-71-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H12Cl2F3N7O2S
Quantity 100 mg
Synonym MLN 2480 BIIB-024
Solubility Information DMSO : 50 mg/mL (98.76 mM; Need ultrasonic)
SMILES NC1=NC=NC(C(N[C@@H](C2=NC=C(C(NC3=NC=C(Cl)C(C(F)(F)F)=C3)=O)S2)C)=O)=C1Cl
Molecular Weight (g/mol) 506.29
Formula Weight 506.29
Percent Purity 98.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.