missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAK-580, MedChemExpress
Description
TAK-580 (MLN 2480) is an orally active and selective inhibitor of pan-Raf kinase.
Specifications
Specifications
| Chemical Name or Material | TAK-580 |
| CAS | 1096708-71-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H12Cl2F3N7O2S |
| Quantity | 100 mg |
| Synonym | MLN 2480 BIIB-024 |
| Solubility Information | DMSO : 50 mg/mL (98.76 mM; Need ultrasonic) |
| SMILES | NC1=NC=NC(C(N[C@@H](C2=NC=C(C(NC3=NC=C(Cl)C(C(F)(F)F)=C3)=O)S2)C)=O)=C1Cl |
| Molecular Weight (g/mol) | 506.29 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?