missing translation for 'onlineSavingsMsg'
Learn More

TAK-220, MedChemExpress

Product Code. 16473917
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473917 50 mg 50mg
16440458 5 mg 5mg
16421909 1 mg 1mg
16417776 100 mg 100mg
16407998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16473917 Supplier Medchem Express Supplier No. HY19974

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TAK-220 is a selective and orally bioavailable CCR5 antagonist, with IC50s of 3.5 nM and 1.4 nM for inhibition on the binding of RANTES and MIP-1α to CCR5, respectively, but shows no effect on the binding to CCR1, CCR2b, CCR3, CCR4, or CCR7

TAK-220 also selectively inhibits HIV-1, with EC50s of 1.2 nM (HIV-1 KK), 0.72 nM (HIV-1 CTV), 1.7 nM (HIV-1 HKW), 1.7 nM (HIV-1 HNK), 0.93 nM (HIV-1

Specifications

Chemical Name or Material TAK-220
CAS 333994-00-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C31H41ClN4O3
Quantity 50 mg
Solubility Information DMSO : ≥ 50 mg/mL (90.39 mM)
SMILES O=C(C1CCN(C(C)=O)CC1)N(CCCN2CCC(CC3=CC=C(C(N)=O)C=C3)CC2)C4=CC=C(C)C(Cl)=C4
Molecular Weight (g/mol) 553.14
Formula Weight 553.14
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.