missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TAK 21d
GREENER_CHOICE

Product Code. 18768589
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18768589

Brand: Tocris Bioscience™ 5209/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent FAAH inhibitor

TAK 21d is a potent FAAH inhibitor; (IC50 values are 0.28 and 0.72 nM, at rat and human FAAH respectively). Displays analgesic effects in vivo models of neuropathic and inflammatory pain. Brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[4-(3,4-Difluorophenyl)-2-pyrimidinyl]-N-3-pyridazinyl-1-piperazinecarboxamide
CAS 1143578-94-2
Quantity 10 mg
Target Fatty Acid Amide Hydrolase (FAAH) Inhibitors
Molecular Formula C19H17F2N7O
Formula Weight 397.38
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.