missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAIP12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102009
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TAIP-12.
The protein encoded by this gene belongs to the CSRNP family of nuclear proteins that share conserved regions, including cysteine- and serine- rich regions, a basic domain, a transcriptional activation domain, and bind the sequence 'AGAGTG', thus have the hallmark of transcription factors. Studies in mice suggest that these genes may have redundant functions. Alternatively spliced transcript variants have been found for this gene.
Specifications
| TAIP12 | |
| Polyclonal | |
| Unconjugated | |
| Csrnp2 | |
| BC035295; C12orf2; C12orf22; Csnrp2; CSRN2; Csrnp2; CSRNP-2; cysteine and serine rich nuclear protein 2; cysteine/serine-rich nuclear protein 2; cysteine-serine-rich nuclear protein 2; Fam130a1; family with sequence similarity 130, member A1; FLJ25576; PPP1R72; Protein FAM130A1; protein phosphatase 1, regulatory subunit 72; RGD1308916; TAIP12; TAIP-12; TGF-beta induced apoptosis protein 12; TGF-beta-induced apoptosis protein 12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 207785, 315308, 81566 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BGQ2, Q9H175 | |
| Csrnp2 | |
| A synthesized peptide derived from human CSRNP2(Accession Q9H175), corresponding to amino acid residues S92-G142. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction