missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAIP12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577271
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The protein encoded by this gene belongs to the CSRNP family of nuclear proteins that share conserved regions, including cysteine- and serine- rich regions, a basic domain, a transcriptional activation domain, and bind the sequence 'AGAGTG', thus have the hallmark of transcription factors. Studies in mice suggest that these genes may have redundant functions. Alternatively spliced transcript variants have been found for this gene.
Specifications
| TAIP12 | |
| Polyclonal | |
| Unconjugated | |
| Csrnp2 | |
| BC035295; C12orf2; C12orf22; Csnrp2; CSRN2; Csrnp2; CSRNP-2; cysteine and serine rich nuclear protein 2; cysteine/serine-rich nuclear protein 2; cysteine-serine-rich nuclear protein 2; Fam130a1; family with sequence similarity 130, member A1; FLJ25576; PPP1R72; Protein FAM130A1; protein phosphatase 1, regulatory subunit 72; RGD1308916; TAIP12; TAIP-12; TGF-beta induced apoptosis protein 12; TGF-beta-induced apoptosis protein 12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 207785, 315308, 81566 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8BGQ2, Q9H175 | |
| Csrnp2 | |
| Synthetic peptide corresponding to amino acids 96-140 of Human TAIP12. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction