missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ISM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA550844
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The antibody detects endogenous levels of total TAIL1 protein.
The protein encoded by this gene contains a type 1 thrombospondin domain, which is present in thrombospondin, a number of proteins involved in the complement pathway, as well as in extracellular matrix proteins. Two alternatively spliced transcript variants encoding distinct isoforms have been observed.
Spezifikation
| ISM2 | |
| Polyclonal | |
| Unconjugated | |
| ISM2 | |
| Gm264; ISM2; isthmin 2; isthmin 2 homolog; isthmin 2 homolog (zebrafish); isthmin-2; PSEC0137; TAIL1; thrombospondin and AMOP containing isthmin-like 1; thrombospondin and AMOP domain-containing isthmin-like protein 1; Thrombospondin type-1 domain-containing protein 3; thrombospondin, type I domain-containing 3; thrombospondin, type I, domain containing 3; THSD3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 145501 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q6H9L7 | |
| ISM2 | |
| Synthetic peptide corresponding to residues near the C terminal of human isthmin 2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur