missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TaiI (MaeII) (10 U/μL)

Product Code. 10121410 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2,000 units
400 units
Unit Size:
2000 units
400 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10121410 400 units 400 units
10795631 2,000 units 2000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10121410 Supplier Thermo Scientific™ Supplier No. ER1141

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cut at ACGT^ sites with TaiI (MaeII) restriction enzyme, which performs best at 65°C in R buffer (Isoschizomers: HpyCH4IV, MaeII).

Lambda DNA digested with TaiI , 1.4% agarose, 143 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme TaiI (MaeII)
Compatible Buffer 10X Buffer R
Sensitive to Heat Inactivation No
Optimal Reaction Temperature 65°C
Type IIS RE No
Quantity 400 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.