missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Clark™ Tags, unstrung, 1A, buff, single, pack of 100
Brand: Fisher Clark™ 4241208
Code : Z4
Additional Details : Weight : 0.71000kg
Description
Unstrung tags for labelling or pricing. Reinforced double-sided washer protects string hole. Made of strong board. Eco-friendly: 100% recycled.- Dimensions: 70mm x 35mm
- Supplied in a pack of 1000