missing translation for 'onlineSavingsMsg'
Learn More

Tafenoquine Succinate, TRC

Product Code. 30416169
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
50 mg
Unit Size:
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416169 10 mg 10mg
30414641 2.5 mg 2.5mg
30411159 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30416169 Supplier Toronto Research Chemicals Supplier No. TRCT00476010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106635-81-8
Molecular Formula C24 H28 F3 N3 O3 . C4 H6 O4
Molecular Weight (g/mol) 581.58
Synonym Butanedioic acid, compd. with N4-[2,6-dimethoxy-4-methyl-5-[3-(trifluoromethyl)phenoxy]-8-quinolinyl]-1,4-pentanediamine (1:1), 1,4-Pentanediamine, N4-[2,6-dimethoxy-4-methyl-5-[3-(trifluoromethyl)phenoxy]-8-quinolinyl]-, butanedioate (1:1) (9CI), 8-[(4-Amino-1-methylbutyl)amino]-2,6-dimethoxy-4-methyl-5-[3-(trifluoromethyl)phenoxy]quinoline succinate, SB 252263, SB 252263AX, Tafenoquine succinate, WR 238605 succinate, SB-252263-AX
IUPAC Name butanedioic acid;4-N-[2,6-dimethoxy-4-methyl-5-[3-(trifluoromethyl)phenoxy]quinolin-8-yl]pentane-1,4-diamine
SMILES COc1cc(C)c2c(Oc3cccc(c3)C(F)(F)F)c(OC)cc(NC(C)CCCN)c2n1.OC(=O)CCC(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H28F3N3O3.C4H6O4/c1-14-11-20(32-4)30-22-18(29-15(2)7-6-10-28)13-19(31-3)23(21(14)22)33-17-9-5-8-16(12-17)24(25,26)27;5-3(6)1-2-4(7)8/h5,8-9,11-13,15,29H,6-7,10,28H2,1-4H3;1-2H2,(H,5,6)(H,7,8)
Formula Weight 581.2349
Chemical Name or Material Tafenoquine Succinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.