missing translation for 'onlineSavingsMsg'
Learn More

Tafenoquine Succinate, MedChemExpress

Product Code. 16467207
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467207 5 mg 5mg
16455356 100 mg 100mg
16450797 50 mg 50mg
16444488 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16467207 Supplier Medchem Express Supplier No. HY111529A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tafenoquine Succinate (WR 238605 Succinate) is an 8-aminoquinoline. Tafenoquine is an anti-malarial prophylactic agent.

Tafenoquine Succinate (WR 238605 Succinate) is an 8-aminoquinoline. Tafenoquine is an anti-malarial prophylactic agent.

Specifications

Chemical Name or Material Tafenoquine Succinate
Physical Form Powder
CAS 106635-81-8
Color Light Nude
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C28H34F3N3O7
For Use With (Application) COVID-19-immunoregulation
Synonym WR 238605 (Succinate)
Solubility Information DMSO : 125 mg/mL (214.93 mM; Need ultrasonic)
SMILES O=C(O)CCC(O)=O.CC(NC1=C2N=C(OC)C=C(C)C2=C(OC3=CC=CC(C(F)(F)F)=C3)C(OC)=C1)CCCN
Molecular Weight (g/mol) 581.58
Formula Weight 581.58
Percent Purity 98.42%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.