missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tafazzin/TAZ Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | Tafazzin/TAZ |
|---|---|
| Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 μL/mg of lysate, Immunohistochemistry-Paraffin 1:5000-1:20000 |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18691436
|
Novus Biologicals
NBP2-98750-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642366
|
Novus Biologicals
NBP2-98750-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tafazzin/TAZ Polyclonal antibody specifically detects Tafazzin/TAZ in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| Tafazzin/TAZ | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Transcription Factors and Regulators | |
| 0.2 um filtered solution in PBS | |
| 6901 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 μL/mg of lysate, Immunohistochemistry-Paraffin 1:5000-1:20000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| BTHSEFE, cardiomyopathy, dilated 3A (X-linked), CMD3A, EFE2FLJ27390, G4.5endocardial fibroelastosis 2, LVNCX, Protein G4.5, tafazzin, Taz1, XAP-2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Tafazzin/TAZ. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title