missing translation for 'onlineSavingsMsg'
Learn More

Tafazzin/TAZ Antibody (3C10), Novus Biologicals™

Product Code. 18323979 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323979 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323979 Supplier Novus Biologicals Supplier No. H00006901M19

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tafazzin/TAZ Monoclonal antibody specifically detects Tafazzin/TAZ in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tafazzin/TAZ
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3C10
Conjugate Unconjugated
Dilution ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH11515
Gene Alias BTHSEFE, cardiomyopathy, dilated 3A (X-linked), CMD3A, EFE2FLJ27390, G4.5endocardial fibroelastosis 2, LVNCX, Protein G4.5, tafazzin, Taz1, XAP-2
Host Species Mouse
Immunogen TAZ (AAH11515, 1 a.a. ~ 262 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPLHVKWPFPAVPPLTWTLASSVVMGLVGTYSCFWTSEWAQAEAGPPGYPCPAGGILKLRHIWNLKLMRWTPAAADICFTKELHSHFFSLGKCVPVCRGDGVYQKGMDFILEKLNHGDWVHIFPEGIGRLIAECHLNPIILPLWHVGEPGDGDREMASGVGGLGLPLVPGCPAPPHVWPSVHCAAGMNDVLPNSPPYFPRFGQKITVLIGKPFSALPVLERLRAENKSAVEMRKALTDFIQEEFQHLKTQAEQLHNHLQPGR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6901
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.