missing translation for 'onlineSavingsMsg'
Learn More

Tafamidis, Mikromol

Product Code. 30462100
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30462100 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462100 Supplier Mikromol Supplier No. MM3971.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Tafamidis, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tafamidis
CAS 594839-88-0
Analyte or Component Names Tafamidis
Shipping Condition Room Temperature
Molecular Formula C14 H7 Cl2 N O3
Certifications/Compliance ISO 17034
Synonym 6-Benzoxazolecarboxylic acid, 2-(3,5-dichlorophenyl)-, 2-(3,5-Dichlorophenyl)-6-benzoxazolecarboxylic acid, Fx 1006, Tafamidis, Vyndaqel
InChI Formula InChI=1S/C14H7Cl2NO3/c15-9-3-8(4-10(16)6-9)13-17-11-2-1-7(14(18)19)5-12(11)20-13/h1-6H,(H,18,19)
Solution Type Neat
SMILES OC(=O)c1ccc2nc(oc2c1)c3cc(Cl)cc(Cl)c3
IUPAC Name 2-(3,5-dichlorophenyl)-1,3-benzoxazole-6-carboxylic acid
Molecular Weight (g/mol) 308.1163
Quantity 250 mg
Formula Weight 306.9803 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.