missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TAFA5/FAM19A5 Antibody (06) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26907MFV450
This item is not returnable.
View return policy
Description
TAFA5/FAM19A5 Monoclonal antibody specifically detects TAFA5/FAM19A5 in Human samples. It is validated for ELISA
Specifications
| TAFA5/FAM19A5 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| Chemokine-like protein TAFA-5, family with sequence similarity 19 (chemokine (C-C motif)-like), member A5, QLLK5208, TAFA protein 5, TAFA5, TAFA-5, UNQ5208 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TAFA5/FAM19A5 (Uniprot#: Q7Z5A7; aa 44-132). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 06 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 25817 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction