missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAFA4/FAM19A4 Antibody (480103), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5099D
This item is not returnable.
View return policy
Description
TAFA4/FAM19A4 Monoclonal antibody specifically detects TAFA4/FAM19A4 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| TAFA4/FAM19A4 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 151647 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 480103 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Chemokine-like protein TAFA-4, family with sequence similarity 19 (chemokine (C-C motif)-like), member A4, FLJ25161, TAFA4, TAFA-4 | |
| E. coli-derived recombinant human TAFA4, Ser35-Arg140, Accession # Q96LR4 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction