missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TAFA1/FAM19A1 Polyclonal antibody specifically detects TAFA1/FAM19A1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Neutralization
Specifications
Specifications
| Antigen | TAFA1/FAM19A1 |
| Applications | Immunohistochemistry, Western Blot, Neutralization |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Chemokine-like protein TAFA-1, family with sequence similarity 19 (chemokine (C-C motif)-like), member A1, TAFA1, TAFA-1 |
| Host Species | Goat |
| Immunogen | E. coli-derived recombinant human TAFA1/FAM19A1, Ser26-Thr133, Accession # NP_998774 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?