missing translation for 'onlineSavingsMsg'
Learn More

TAFA1/FAM19A1 Antibody [Allophycocyanin], Novus Biologicals™

Product Code. 30738366 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738366 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738366 Supplier Novus Biologicals Supplier No. AF5154A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

TAFA1/FAM19A1 Polyclonal antibody specifically detects TAFA1/FAM19A1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAFA1/FAM19A1
Applications Immunohistochemistry, Western Blot, Neutralization
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias Chemokine-like protein TAFA-1, family with sequence similarity 19 (chemokine (C-C motif)-like), member A1, TAFA1, TAFA-1
Host Species Goat
Immunogen E. coli-derived recombinant human TAFA1/FAM19A1, Ser26-Thr133, Accession # NP_998774
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 407738
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.