missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAF9B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103026
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TAF9B.
The protein encoded by this gene is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine. The encoded protein is involved in myelopoiesis, and defects in this gene have been associated with several myeloproliferative disorders. Two variants encoding different isoforms have been found for this gene.
Specifications
| TAF9B | |
| Polyclonal | |
| Unconjugated | |
| Taf9b | |
| BC066223; Dn7; DN-7; neuronal cell death-related gene in neuron 7; neuronal cell death-related gene in neuron-7; neuronal cell death-related protein 7; Taf9b; TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31kDa; TAF9L; TAF9-like RNA polymerase II TATA box binding protein (TBP)-associated factor 31 kD; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31 kD; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31kDa; TAFII31; TAFII31L; TATA box binding protein associated factor 9b; TATA-box binding protein associated factor 9b; TBP-associated factor 9L; TFIID-31; transcription associated factor TAFII31L; transcription initiation factor IID, 31kD subunit; Transcription initiation factor TFIID subunit 9B; Transcription initiation factor TFIID subunit 9-like; transcription-associated factor TAFII31L | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 407786, 51616 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6NZA9, Q9HBM6 | |
| Taf9b | |
| A synthesized peptide derived from human TAF9B(Accession Q9HBM6), corresponding to amino acid residues M1-Q38. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction