missing translation for 'onlineSavingsMsg'
Learn More

TAF9b Antibody, Novus Biologicals™

Product Code. 18377259 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377259 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377259 Supplier Novus Biologicals Supplier No. H00051616D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TAF9b Polyclonal antibody specifically detects TAF9b in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF9b
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_057059.2
Gene Alias 31kDa, DN7, DN-7Transcription-associated factor TAFII31L, Neuronal cell death-related protein 7, TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF9Ltranscription associated factor TAFII31L, TAFII31LTBP-associated factor 9L, TFIID-31, transcription initiation factor IID, 31kD subunit, transcription initiation factor TFIID subunit 9B, Transcription initiation factor TFIID subunit 9-like
Host Species Rabbit
Immunogen TAF9B (NP_057059.2, 1 a.a. - 251 a.a.) full-length human protein. MESGKMAPPKNAPRDALVMAQILKDMGITEYEPRVINQMLEFAFRYVTTILDDAKIYSSHAKKPNVDADDVRLAIQCRADQSFTSPPPRDFLLDIARQKNQTPLPLIKPYAGPRLPPDRYCLTAPNYRLKSLIKKGPNQGRLVPRLSVGAVSSKPTTPTIATPQTVSVPNKVATPMSVTSQRFTVQIPPSQSTPVKPVPATTAVQNVLINPSMIGPKNILITTNMVSSQNTANEANPLKRKHEDDDDNDIM
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51616
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.