missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF9b Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94264-0.02ml
This item is not returnable.
View return policy
Description
TAF9b Polyclonal antibody specifically detects TAF9b in Human samples. It is validated for Western Blot
Specifications
| TAF9b | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 31kDa, DN7, DN-7Transcription-associated factor TAFII31L, Neuronal cell death-related protein 7, TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF9Ltranscription associated factor TAFII31L, TAFII31LTBP-associated factor 9L, TFIID-31, transcription initiation factor IID, 31kD subunit, transcription initiation factor TFIID subunit 9B, Transcription initiation factor TFIID subunit 9-like | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human TAF9B (NP_057059.2). ILDDAKIYSSHAKKPNVDADDVRLAIQCRADQSFTSPPPRDFLLDIARQKNQTPLPLIKPYAGPRLPPDRYCLTAPNYRLKSLIKKGPNQGRLVPRLSVGA | |
| 0.02 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51616 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction