missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAF9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110457
This item is not returnable.
View return policy
Description
Immunogen sequence: MESGKTASPK SMPKDAQMMA QILKDMGITE YEPRVINQML EFAFRYVTTI LDDAKIYSSH AKKATVDADD VRLAIQCRAD QSFTSPPPRD FLLDIARQRN QTPLPLIKPY SGPRLPPDRY CLTAPNYRLK SLQKKASTSA GRITVPRLSV GSVTSRPSTP TLGTPTPQTM SV.
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the smaller subunits of TFIID that binds to the basal transcription factor GTF2B as well as to several transcriptional activators such as p53 and VP16. A similar but distinct gene (TAF9L) has been found on the X chromosome and a pseudogene has been identified on chromosome 19. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| TAF9 | |
| Polyclonal | |
| Unconjugated | |
| TAF9 | |
| 2310012M09Rik; AA673500; AD-004; AU045423; CGI-137; CINAP; CIP; EMBL:AAH91109.1}; hCINAP; MGC:1603; MGC:3647; MGC:5067; RNA polymerase II TBP-associated factor subunit G; STAF31/32; TAF2G; TAF9; taf9 {ECO:0000312; TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 32 kDa; TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 32kDa; TAF9 RNA polymerase II, TATA box binding protein-associated factor; Tafii31; TAFII-31; TAFII32; TAFII-32; TAFIID32; TATA box binding protein associated factor 9; TATA-box binding protein associated factor 9; transcription initiation factor TFIID 31 kD subunit; transcription initiation factor TFIID 31 kDa subunit; Transcription initiation factor TFIID 32 kDa subunit; transcription initiation factor TFIID subunit 9 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 373541, 6880 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16594, Q5BKE0 | |
| TAF9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-172 of human TAF9 (NP_0031781). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction