missing translation for 'onlineSavingsMsg'
Learn More

TAF8 Antibody, Novus Biologicals™

Product Code. 18414089 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414089 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414089 Supplier Novus Biologicals Supplier No. H00129685B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

TAF8 Polyclonal antibody specifically detects TAF8 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF8
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Alias 45/50kDa, FLJ32821, II, Protein taube nuss, RNA polymerase II, 43 kD, TAF(II)43,43, TAF8 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 43kDa, TATA box binding protein (TBP)-associated factor, RNA polymerase II, A, taube nuss homolog (mouse), TBP-associated factor 43 kDa, TBP-associated factor 8, transcription initiation factor TFIID subunit 8
Host Species Mouse
Immunogen TBN (AAH33728.1, 1 a.a. - 174 a.a.) full-length human protein. MADAAATAGAGGSGTRSGSKQSTNPADNYHLARRRTLQVVVSSLLTEAGFESAEKASVETLTEMLQSYISEIGRSAKSYCEHTARTQPTLSDIVVTLVEMGFNVDTLPAYAKRSQRMVITAPPVTNQPVTPKALTAGQNRPHPPHIPSHFPEFPDPHTYIKTPVSDEALGLRVV
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129685
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.