missing translation for 'onlineSavingsMsg'
Learn More

TAF7L Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18319402 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319402 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319402 Supplier Novus Biologicals Supplier No. NBP306642AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TAF7L Polyclonal antibody specifically detects TAF7L in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF7L
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias 50kDa, CT40FLJ23157, RNA polymerase II TBP-associated factor subunit Q, TAF2QRNA polymerase II, Q, TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, TATA box binding protein (TBP)-associated factor, RNA polymerase II, Q, TATA box-binding protein-associated factor 50 kDa, transcription initiation factor TFIID subunit 7-like
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TAF7L fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54457
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.