Learn More
Invitrogen™ TAF7 Monoclonal Antibody (19TA-2C7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA3079
Description
MA3-079 detects TATA-Binding Protein (TBP) Associated factor 7 from human, canine and non-human primate samples. MA3-079 has been successfully used in Western blot, immunofluorescence and flow cytometry applications. The MA3-079 immunogen is a recombinant human TAF7 protein.
The intronless gene for this transcription coactivator is located between the protocadherin beta and gamma gene clusters on chromosome 5. The protein encoded by this gene is a component of the TFIID protein complex, a complex which binds to the TATA box in class II promoters and recruits RNA polymerase II and other factors. This particular subunit interacts with the largest TFIID subunit, as well as multiple transcription activators. The protein is required for transcription by promoters targeted by RNA polymerase II. The general transcription factor, TFIID, consists of the TATA-binding protein (TBP) associated with a series of TBP-associated factors (TAFs) that together participate in the assembly of the transcription preinitiation complex.
Specifications
| TAF7 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.05% sodium azide | |
| Q15545 | |
| TAF7 | |
| Recombinant human TAF7 protein. | |
| RUO | |
| 487180, 6879 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 19TA-2C7 | |
| Unconjugated | |
| TAF7 | |
| 55kDa; AI607308; BB007175; RNA polymerase II TBP-associated factor subunit F; TAF(II)55; TAF2F; Taf7; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55 kDa; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa; TAFII55; TAFII-55; TATA box binding protein (TBP) associated factor, RNA polymerase II, F, 55 kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, F, 55kD; TATA box binding protein associated factor 7; TATA-box binding protein associated factor 7; TBP-associated factor F; transcription factor IID subunit TAFII55; transcription initiation factor TFIID 55 kDa subunit; Transcription initiation factor TFIID subunit 7; transcription initiation factor TFIID, 55 kDa subunit | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human, Canine, Monkey | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.